Oakton® pH Testr™ 50 Waterproof Pocket pH Tester, Premium 50 Series, Cole-Parmer
Supplier: Argos Technologies
35634-15
76208-088EA 413.97 USD InStock
76208-088
Oakton® pH Testr™ 50 Waterproof Pocket pH Tester, Premium 50 Series, Cole-Parmer
Electrochemistry Meters pH and Multi-Parameter Meters
Versatile, compact design works in the field or laboratory.
  • Easy-to-navigate interface for quick setup and calibration
  • Replaceable, double-junction electrode sensors save time and money
  • Automatic temperature compensation ensures accurate reading in varying environments
  • Waterproof
  • LCD display
Redesigned leakproof cap functions as a sample cup, hydrating cup and stabilizing base to stand the meter upright.
Información para pedidos: Includes protective plastic storage case, lanyard, and 4 AAA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty