QuantiQuik™ Pyruvate Quick Test Strips, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
QQPYR10
76234-760EA 110.59 USD InStock
76234-760
QuantiQuik™ Pyruvate Quick Test Strips, BioAssay Systems
Assays Cellular Assays
Direct determination of Pyruvate concentrations in food and beverage samples as well as biological samples.
  • Fast and sensitive
  • Use of 50, 100, or 400 µl sample
  • Semi-quantitative measurement between 0-131 mg/l (undiluted) Pyruvate
  • Convenient. No expensive lab equipment needed
  • Sample treatment and assay can be performed in under 15 minutes
  • Method: Visual
  • Samples: Wine, beer, biological samples, and more
Pyruvate, a key intermediate in cellular metabolic pathways, can be converted to a variety of things such as carbohydrates, fatty acids, and energy depending on the pathway. It can also be converted to alanine and ethanol. In the beverage industry, monitoring levels of pyruvate during alcoholic fermentation can be important for production of beer, wine, and spirits. Some brewers even add sodium pyruvate to their beer to alter the flavor and give the beer more body. In regards to health, abnormal levels of pyruvate have been linked to various liver diseases and metabolic disorders.
QuantiQuik™ Pyruvate Test Strips are based on pyruvate oxidase catalyzed oxidation of pyruvate in which peroxidase reduces the formed peroxide and oxidizes a chromogenic reagent. The intensity of the blue product is directly proportional to pyruvate concentration in the sample.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty