Photochemistry Immersion Lamps, Ace Glass Incorporated
Supplier: Ace Glass
7825-35 7825-40 7825-30 7825-32 7825-34 7825-36 7825-300 7825-320 7825-340 7825-350 7825-400
89084-182EA 1706.87 USD InStock
89084-182 89084-184 89084-186 89084-188 89084-190 89084-192 76318-560 76318-562 76318-564 76318-566 76318-570
Photochemistry Immersion Lamps, Ace Glass Incorporated
Lights Mercury Lamps
Medium pressure, quartz, mercury-vapor lamp
For use in all Ace Glass immersion wells. 61 cm PTFE covered lead wires, fitted with pin type connectors. A six-foot power cord also allows for lowering the lamp into the well for vertical operation. Approximately 40 to 48% of the radiated energy is in the ultraviolet portion of the spectrum; 40 to 43% in the visible region; and the balance in the infrared. Can be inserted into a glass well/sleeve with a 25 mm I.D
Información para pedidos: CAUTION–Ultra-violet radiation is permanently damaging to the retina of the eye. Never operate this lampMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty