VWR® Phenolic Bottle Caps, Black
Supplier: VWR International
10862-216 10862-218 10862-220 10862-222 10862-224 10862-270 10862-272 10862-274 10862-276 10862-278 10862-280 10862-282 10862-284 10862-286 10862-288 10862-290 10862-292 10862-294 10862-296 10862-298 10862-300 10862-302 10862-388 10862-390 10862-400 10862-408 10862-410 10862-412 10862-418 10862-422 10862-424 10862-426 10862-432 10862-434 10862-436 10862-438 10862-440 10862-442 10862-446 10862-448 10862-452 10862-454 10862-456 10862-460 10862-462 10862-464 10862-468 10862-472 10862-474 10862-476 10862-478 10862-480 10862-484 76329-632 76329-636 76329-638 76329-640 76329-642 76329-644 76329-646
10862-216CS 47.92 USD InStock
10862-216 10862-218 10862-220 10862-222 10862-224 10862-270 10862-272 10862-274 10862-276 10862-278 10862-280 10862-282 10862-284 10862-286 10862-288 10862-290 10862-292 10862-294 10862-296 10862-298 10862-300 10862-302 10862-388 10862-390 10862-400 10862-408 10862-410 10862-412 10862-418 10862-422 10862-424 10862-426 10862-432 10862-434 10862-436 10862-438 10862-440 10862-442 10862-446 10862-448 10862-452 10862-454 10862-456 10862-460 10862-462 10862-464 10862-468 10862-472 10862-474 10862-476 10862-478 10862-480 10862-484 76329-632 76329-636 76329-638 76329-640 76329-642 76329-644 76329-646
VWR® Phenolic Bottle Caps, Black
Closures Bottle Closures
VWR® Black Phenolic Caps provide a wide range of chemical compatibility and temperature tolerance with an assortment of liners to choose from.
  • Available with a variety of liners to meet specific requirements
  • Match the bottle or jar neck size to the cap size to find the perfect fit
Pulp/Vinyl general purpose liners are commonly used for food, beverage, medical and chemical applications packed at less than 120 °F, including mild acids, alkalis, solvents, alcohols, oils and aqueous products; not recommended for active hydrocarbons or bleaches.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty