Male Luer Lock, Needleless Injection Site, for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1399-F50
76333-462PK 113.95 USD InStock
76333-462
Male Luer Lock, Needleless Injection Site, for Bioreactors, Chemglass
Fittings and Connectors
Male Luer lock with needleless injection site and swabable polycarbonate, silicone with lach samplers for bioreactors.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty