G460 Multi-gas Detectors, GfG Instrumentation
Supplier: GFG INSTRUMENTATION, INC.
4600D03706420 4601D03100020 4601D03100065 4601D03100065K 4601D03200020 4601D03240020 4601D03240065 4601D03240065K 4601D03700020 4601D03700065 4601D03700065K
76334-702EA 6985.28 USD InStock
76334-702 76334-704 76334-706 76334-708 76334-710 76334-712 76334-714 76334-716 76334-718 76334-720 76334-722 76334-724 76334-726
G460 Multi-gas Detectors, GfG Instrumentation
Gas Detection Systems
The G460 is a rugged, compact instrument for detection of up to six gases. The concussion proof housing, along with the dust and water resistant housing protects the instrument in the harshest environments. It features completely automatic calibration, one-button operation, top mounted display and interchangeable battery packs for up to 24 hours of operation. The rechargeable battery pack can even be equipped with optional 'flash light' LEDs.
  • Exceptional HAZMAT and confined space performance
  • Up to six channels of detection
  • Optional PID sensor for direct
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty