ZD 21 Series Fixed Transmitter, Fixed Systems Gas Detection, GfG Instrumentation
Supplier: GFG INSTRUMENTATION, INC.
2210009 2210404
76334-866EA 995.12 USD InStock
76334-866 76335-132
ZD 21 Series Fixed Transmitter, Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
The ZD 21 is an “intelligent” transmitter. The comprehensive electronics allow for easy operation and maintenance and also increase operational safety and accuracy. In less than 2 seconds the ZD 21 responds to an oxygen deficiency. This extremely quick response time allows the time to take measures to prevent considerable damage.
  • Extremely stable sensor responds quickly to any change in oxygen (O₂) concentration
  • Sensor is unaffected by exposure to high concentrations of carbon dioxide (CO₂)
  • 5 year oxygen (O₂) sensor life (typical)
  • Fast recovery from
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty