Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific

Supplier: VWR
CRTPR011WWG/0 CRTPR011WWGLH/0 CRTPR011WWW/0 CRTPR011WWWLH/0 CRTPR031WWG/0 CRTPR031WWW/0 CRTPR051WWG-0 CRTPR051WWGLH0 CRTPR051WWW-0 CRTPR051WWWLH0 CRTPR061WWG-0 CRTPR101WWG-0 CRTPR101WWW-0
76336-666EA 3204.73 USD InStock
76336-666 76336-682 76336-684 76336-686 76336-688 76336-690 76336-692 76336-694 76336-696 76336-698 76336-700 76336-702 76336-704
Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific
Testing Chambers Climatic Chambers
These CRT cabinets provide controlled room temperature environments for storing sensitive substances that require precise temperature management.

  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft air circulation
  • Probe access
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*

These specialized cabinets are ideal for scientific and medical applications. Both built-in and freestanding configurations are available. Built-in units are designed with front-facing ventilation systems, allowing easyMore Product Information
Order Now
 

ORDER

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Description Volume No. of shelves Ext. dimensions Electrical Shipping weight Supplier No. VWR Catalog Number Unit Price Quantity
Countertop controlled room temperature cabinet freestanding, glass door 28.3 L (1 cu. ft.) 2 adjustable 171/4W×191/4D×211/4" H 115 V, 0.9 A 41.3 kg (91 lbs.) CRTPR011WWG/0 76336-666 Each
Retrieving
Safer Human and Environmental Health

Countertop controlled room temperature cabinet freestanding, glass door, left hinged 28.3 L (1 cu. ft.) 2 adjustable 171/4W×191/4D×211/4" H 115 V, 0.9 A 41.3 kg (91 lbs.) CRTPR011WWGLH/0 76336-682 Each
Retrieving
Safer Human and Environmental Health

Countertop controlled room temperature cabinet freestanding, solid door 28.3 L (1 cu. ft.) 2 adjustable 171/4W×191/4D×211/4" H 115 V, 0.9 A 41.3 kg (91 lbs.) CRTPR011WWW/0 76336-684 Each
Retrieving
Safer Human and Environmental Health

Countertop controlled room temperature cabinet freestanding, solid door, left hinged 28.3 L (1 cu. ft.) 2 adjustable 171/4W×191/4D×211/4" H 115 V, 0.9 A 41.3 kg (91 lbs.) CRTPR011WWWLH/0 76336-686 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet freestanding, glass door, ADA compliant 70.8 L (2.5 cu. ft.) 4 adjustable 173/4W×191/2D×287/8" H 115 V, 0.9 A 44 kg (97 lbs.) CRTPR031WWG/0 76336-688 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet freestanding, solid door, ADA compliant 70.8 l (2.5 cu.ft.) 4 adjustable 173/4W×191/2D×287/8" H 115 V, 0.9 A 38.5 kg (85 lbs.) CRTPR031WWW/0 76336-690 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet built-in, glass door 130.3 L (4.6 cu. ft.) 3 (2 adjustable/1 fixed) 233/4W×251/2D×333/8" H 115 V, 1.74 A 68 kg (150 lbs.) CRTPR051WWG-0 76336-692 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet built-in, glass door, left hinged 130.3 L (4.6 cu. ft.) 3 (2 adjustable/1 fixed) 233/4W×251/2D×333/8" H 115 V, 1.74 A 68 kg (150 lbs.) CRTPR051WWGLH0 76336-694 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet built-in, solid door 130.3 L (4.6 cu. ft.) 3 (2 adjustable/1 fixed) 233/4W×241/2D×333/8" H 115 V, 1.74 A 62 kg (137 lbs.) CRTPR051WWW-0 76336-696 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet built-in, solid door, left hinged 130.3 L (4.6 cu. ft.) 3 (2 adjustable/1 fixed) 233/4W×241/2D×333/8" H 115 V, 1.74 A 62 kg (137 lbs.) CRTPR051WWWLH0 76336-698 Each
Retrieving
Safer Human and Environmental Health

Undercounter controlled room temperature cabinet freestanding, glass door 147.2 L (5.2 cu. ft.) 3 adjustable 233/4W×24D×321/8" H 115 V, 1.3 A 58.5 kg (129 lbs.) CRTPR061WWG-0 76336-700 Each
Retrieving
Safer Human and Environmental Health

Upright controlled room temperature cabinet, glass door 297.3 L (10.5 cu. ft.) 6 adjustable 233/4W×261/2D×59" H 115 V, 2.30 A 94.8 kg (209 lbs.) CRTPR101WWG-0 76336-702 Each
Retrieving
Safer Human and Environmental Health

Upright controlled room temperature cabinet, solid door 297.3 L (10.5 cu. ft.) 6 adjustable 233/4W×261/2D×59" H 115 V, 2.30 A 83 kg (183 lbs.) CRTPR101WWW-0 76336-704 Each
Retrieving
Safer Human and Environmental Health


Generate Barcodes
Generate PDF Catalog Page
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

SPECIFICATIONS

ACCESSORIES

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty