Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific
Supplier: VWR
CRTPR011WWG/0 CRTPR011WWGLH/0 CRTPR011WWW/0 CRTPR011WWWLH/0 CRTPR031WWG/0 CRTPR031WWW/0 CRTPR051WWG-0 CRTPR051WWGLH0 CRTPR051WWW-0 CRTPR051WWWLH0 CRTPR061WWG-0 CRTPR101WWG-0 CRTPR101WWW-0
76336-666EA 3204.73 USD InStock
76336-666 76336-682 76336-684 76336-686 76336-688 76336-690 76336-692 76336-694 76336-696 76336-698 76336-700 76336-702 76336-704
Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific
Testing Chambers Climatic Chambers
These CRT cabinets provide controlled room temperature environments for storing sensitive substances that require precise temperature management.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft air circulation
  • Probe access
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These specialized cabinets are ideal for scientific and medical applications. Both built-in and freestanding configurations are available. Built-in units are designed with front-facing ventilation systems, allowing easyMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty