Monarch® gDNA Tissue Lysis Buffer, New England Biolabs
Supplier: W.u.G. Dickgießer GmbH
T3011L
76339-774EA 52 USD InStock
76339-774
Monarch® gDNA Tissue Lysis Buffer, New England Biolabs
Buffers Extraction Buffers
The Monarch® gDNA Tissue Lysis Buffer is a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample types.
The Tissue Lysis Buffer supplies intermediate lysis conditions that enable rapid digestion of the tissue pieces while simultaneously ensuring the genomic DNA fragment length is optimal for binding and elution.
The Monarch® gDNA Tissue Lysis Buffer is one of three lysis buffers available for purchase separately for additional convenience and to minimize waste.
Coupled with optimized Proteinase K digestion, the tissue lysis system facilitates above-average yields for common animal tissues , including tissues that prove difficult for many other commercial kits (e.g., brain and muscle).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty