Heracell i CO₂ Incubators with Variable O₂ Control, Thermo Scientific
Supplier: Thermo Scientific
51026410 51026408 51026402 51026537 51026529 51026423 51026422 51026556 51026533 51032967 51032969 51032971 51032972 51032973 51032975 51032977
89187-188EA 16500 USD InStock
89187-188 89187-190 89187-192 89187-194 89187-196 89187-200 89187-202 89187-204 89187-206 76379-706 76379-708 76379-710 76379-712 76379-714 76379-716 76379-720
Heracell i CO₂ Incubators with Variable O₂ Control, Thermo Scientific
Incubators CO₂ Incubators
Heracell i CO₂ incubators with variable control of oxygen stimulate cell behaviors that are more predictive of the in vivo environment. Available in a choice of two ranges of O₂ control, 1 to 21% for physiological levels and 5 to 90% for hyperoxic studies.each is supplied with maintenance free zirconia oxide sensor is calibrated automatically and remains in place during the ContraCon high temperature decontamination, and a unique gas tight segmented inner door assembly enables access sections of the chamber without disturbing the entire environment to minimize recovery time, contaminationMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICIOS
Servicios: Thermo Scientific provides several service options such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Heracell CO2 Incubator Technical Services" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty