ASTM Individual Calibration Weights and Calibration Weight Sets, Troemner
Supplier: Troemner
30390136 30390137 30390138 30390139 30390140 30390141 30390143 30390144 30390145 30390148 80859285 30390150 30390153 30390017 30390155 80858689 80858651 80858635 80858599 80858558 80858519 80858503 80858470 80858431 80858394 80858358 80858318 80858299 80858284 80858271 80858254 80858239 80858225 80858211
76404-580EA 1479.57 USD InStock
76404-580 76404-582 76404-584 76404-586 76404-588 76404-590 76404-592 76404-594 76404-596 76404-598 76404-600 76404-602 76404-604 76404-606 76404-608 76404-610 76404-612 76404-614 76404-616 76404-618 76404-620 76404-622 76404-624 76404-626 76404-628 76404-630 76404-632 76404-634 76404-636 76404-638 76404-640 76404-642 76404-644 76404-646
ASTM Individual Calibration Weights and Calibration Weight Sets, Troemner
Weights Calibration Weights
Alloy 8, high grade stainless steel calibration weights and reference standards. NVLAP, NIST Traceable calibration certificates in UltraClass Series and ANSI/ASTM Class 000, 00, 0, 1, 2, 3, and 4.
  • Range: 1 mg to 50 kg, 1 mg to 500 mg
  • Style: Cylindrical, flat, wire
  • Density: 1 g and above: 8.03 g/cm³; 500 mg to 5 mg: 7.95 g/cm³; 5 mg and below: 2.70 g/cm³
Weights and weight sets are manufactured with the finest stainless steels available, including Troemner's proprietary Alloy 8. Specification on surface finish, markings, construction, material density,More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty