VWR® Certified Reference Materials, Oxide
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
21001491
76411-358CS 1278.2 USD InStock
76411-358
VWR® Certified Reference Materials, Oxide
Standards Cationic Standards Gold Standards
KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels
Información para pedidos: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty