EasyReach™ Cleaning Tools, Contec®
Supplier: Contec
MEQT0001 PSME0001 2685 2685LF 2665LF MEQT0021 MEQT0022 MEQT0002SZ 2685P
89094-578CS 362.83 USD InStock
89094-578 89094-580 89094-582 89094-584 89107-994 76417-120 76417-122 76417-132 76417-134 76449-778 76480-378
EasyReach™ Cleaning Tools, Contec®
Mops
The lightweight, stainless steel EasyReach™ Cleaning Tool is designed for cleaning isolators, laminar air flow hoods, glove boxes, and other mini environments. Rectangular shape makes overlapping straight-line action easy to achieve. Hinged joint and lightweight design makes one-handed cleaning comfortable and improves access to hard-to-reach areas.
  • Rounded tool edges and strong, durable, double-layer pads ensure equipment is not damaged during use
  • Lightweight EasyReach™ Tool can easily be manipulated, is comfortable to use, and minimizes fatigue
  • All
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty