Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Safety Poster™ Sign, 'HAND HYGIENE' (Bilingual English/Spanish) Accuform®

Supplier: ACCUFORM MANUFACTURING, INC
SP124486 SP124486L SP124486J SP124486JL
76442-652EA 21.45 USD InStock
76442-652 76442-654 76442-656 76442-658
Safety Poster™ Sign, 'HAND HYGIENE' (Bilingual English/Spanish) Accuform®
Signs
Safety Posters communicate the importance of safety procedures and practices throughout the workplace.

  • Multiple material and size options available

Choose from two material options. Non-Laminated Posters are printed with bright, colorful words and graphics. They are an economical solution for indoor use. Laminated Posters are also printed with bright, colorful words and graphics. They are laminated on both sides, and designed for tough, industrial-type environments.

Order Now
 

ORDER


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Dimensions Material Legend Supplier No. VWR Catalog Number Unit Price Quantity
22×17" Non-laminated HAND HYGIENE (Bilingual English/Spanish) SP124486 76442-652 Each
Retrieving
22×17" Laminated HAND HYGIENE (Bilingual English/Spanish) SP124486L 76442-654 Each
Retrieving
28×22" Non-laminated HAND HYGIENE (Bilingual English/Spanish) SP124486J 76442-656 Each
Retrieving
28×22" Laminated HAND HYGIENE (Bilingual English/Spanish) SP124486JL 76442-658 Each
Retrieving

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty