Heracell VIOS 250i CO₂ Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
51033586 51033587 51033596 51033597 51033800 51033804 51033806 51033807 51033783 51033786
76459-174EA 20200 USD InStock
76459-174 76459-176 76459-178 76459-180 76459-202 76459-204 76459-206 76459-208 76529-598 76529-600
Heracell VIOS 250i CO₂ Incubators, Thermo Scientific
Incubators CO₂ Incubators
Designed to achieve your next breakthrough. Better solutions for optimal cell growth, VIOS CO₂ incubators provide the ideal in-vitro environment.
  • Now you can choose between the 165L or the 255L capacity
  • Pick the CO₂ incubator that's right for your lab's needs
  • Ideal for scale-up research and development
  • 255L chamber accommodates high throughput and large culture vessels
  • Strengthened stainless steel models for increased weight capacity
  • Optional reinforced shelves for large capacity, low media level culturing
The Heracell VIOS CO₂ incubator delivers theMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICIOS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty