Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azure B (azure I), powder_dark green, certified C.I. 52010

Supplier: E. Becker & Co. GmbH

0219052010 0219052001
IC19052010EA 137.28 USD InStock
IC19052010 76461-470
Azure B (azure I), powder_dark green, certified C.I. 52010
Azure B (azure I)
Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.

Soluble in water (50 mg/ml) or ethanol (8-10 mg/ml); insoluble in diethyl ether.

Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.


Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Melting Pt: 205…210 °C (dec., lit.)
MDL Number: MFCD00011935
CAS Number: 531-55-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbf74f5af7111683a65da1ac546c907ea65520c&vid=bdbf74f5af7111683a65da1ac546c907ea65520c&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty