Vacuum Jacketed Distillation Column, Ace Glass
Supplier: Ace Glass
6602-02 6602-03
76470-468EA 955.8 USD InStock
76470-468 76470-470
Vacuum Jacketed Distillation Column, Ace Glass
Distillation Components Distillation Columns
Vacuum jacketed, straight thru column, when mated to 76470 family of condensers, completes a short path distillation head.
  • Two-piece design simplies cleaning
  • Ball and socket joint allow for adjustability
  • High-quality borosilicate glass construction
Two-piece design simplifies cleaning and our 35/25 O-ring ball and socket joint connection allows for some adjustability over a standard taper joint configuration. For use with or without customer supplied packing material.
Información para pedidos: For a complete short path distillation head order with 7More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty