VWR® DuraChill® Recirculating Chillers, -10 to 70 °C, 240 V
Supplier: VWR International
76472-552 76472-554 76472-560 76472-562 76472-568 76472-570 76472-576 76472-578
76472-552EA 7106.81 USD InStock
76472-552 76472-554 76472-560 76472-562 76472-568 76472-570 76472-576 76472-578
VWR® DuraChill® Recirculating Chillers, -10 to 70 °C, 240 V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
DuraChill® chillers provide stable and reliable cooling for various common heat removal applications, including laser etching, AA furnaces, ICP, rotary evaporators, vacuum systems, reaction vessels, plasma etching, and condenser cooling.
  • Large, full-color touchscreen display with intuitive interface and support for five different languages
  • Compact, portable design takes up less floor space
  • DynamicFilter™ self-changing filter system
  • Front fill reservoir
  • Liquid level monitoring to prevent pump failure
  • Ultraviolet anti-biological light system
  • User-adjustable
More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty