Single-Sample Cryoscope, Model 4250, Advanced Instruments
Supplier: Advanced Instruments
4250
76474-142EA 20658.5 USD InStock
76474-142
Single-Sample Cryoscope, Model 4250, Advanced Instruments
Cryoscopes
The single-sample cryoscope, model 4250 is a fast, reliable, and precise system for milk analysis.
  • Low maintenance
  • On-board printer for better traceability
  • Rapid and stable automated calibration
The model 4250 employs the industry preferred freezing point depression (FPD) method for determination of added water to milk. It allows your plant to manage a premium milk supply, ensure quality products and improve your operating efficiency.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty