Ptec™ Bench Top Ionizer, Removable Emitter Assembly, Transforming Technologies
Supplier: Transforming Technologies
IN2000
76474-664EA 977.08 USD InStock
76474-664
Ptec™ Bench Top Ionizer, Removable Emitter Assembly, Transforming Technologies
Ionizers
The Ptec™ IN2000 AC bench top air ionizer is a small, efficient static elimination device with variable fan speed for protecting electronic components and general static protection.
  • Remove static charges quickly and efficiently
  • Removable emitter point system simplifies service and maintenance
  • Perfect for single station table top, bench, or desk use
  • Balance: ±15 V
  • Small footprint
The IN2000 emitter assembly can be quickly removed and cleaned which eliminates maintenance costs and equipment downtime and ensures the ionizer has optimal static eliminatingMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty