Amplite® Colorimetric Glucose Oxidase Assay Kit , AAT Bioquest
Supplier: AAT BIOQUEST INC
11299
76481-900EA 375.27 USD InStock
76481-900
Amplite® Colorimetric Glucose Oxidase Assay Kit , AAT Bioquest
Assays Enzyme Assays
The glucose oxidase is a dimeric protein that catalyzes the oxidation of beta-D-glucose into hydrogen peroxide and D-glucono-1,5-lactone, which is hydrolyzed to gluconic acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty