Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Resorufin alpha-D-galactopyranoside

Supplier: AAT BIOQUEST INC
Warning

14021
76482-302EA 375.27 USD InStock
76482-302
Resorufin alpha-D-galactopyranoside
Resorufin alpha-D-galactopyranoside
Resorufin alpha-D-galactopyranoside is a sensitive fluorogenic substrate that generates a red fluorescent product (resorufin) upon interaction with alpha-galactosidase.

  • Alpha-galactosidase fluorogenic substrate
  • Sensitive detection
  • Multiple applications

Precaución: For research use only


MW: 375.33 g/mol
ADR: 5 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty