DAPI Dilactate 95
Supplier: AAT BIOQUEST INC
17509
76482-848EA 124.31 USD InStock
76482-848
DAPI Dilactate 95
DAPI Dilactate
DAPI is a fluorescent stain that binds strongly to DNA.
  • Binding-Determined Bicolor Fluorescence
  • Cell-Permeable
  • Useful For Multicolor Imaging
Precaución: For Research Use Only
CAS Number: 28718-91-4
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty