Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DiSBAC2(3) measurement_membrane potential

Supplier: AAT BIOQUEST INC
Warning

21414
76483-388EA 124.31 USD InStock
76483-388
DiSBAC2(3) measurement_membrane potential
DiSBAC2(3)


Formula: C₁₉H₂₄N₄O₄S₂
MW: 436.55 g/mol
CAS Number: 47623-98-3
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad3dd08a4e1e770b5f2a247b90b0f1d493ef86dd&vid=ad3dd08a4e1e770b5f2a247b90b0f1d493ef86dd&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a12ea7694ac91e8c85171b9195572d1e389e567e&vid=a12ea7694ac91e8c85171b9195572d1e389e567e&prid=web&clid=SAPDAM
Order Now

 

ORDER


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Supplier No. VWR Catalog Number Unit Price Quantity
21414 76483-388 Each (25mg)
Retrieving
Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.


restricted This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty