Cell Explorer™ Live Cell Tracking Kit *Orange Fluorescence*, AAT Bioquest
Supplier: AAT BIOQUEST INC
22622
76483-634EA 249.58 USD InStock
76483-634
Cell Explorer™ Live Cell Tracking Kit *Orange Fluorescence*, AAT Bioquest
Assays Cellular Assays
Our Cell Explorer™ Live Cell Tracking Kit uses a proprietary orange fluorescent dye that gets enhanced fluorescence upon entering into live cells.
  • Long label retention
  • Suitable for all major fluorescence platforms
  • Contains all necessary components
  • Multiple applications
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty