Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*, AAT Bioquest
Supplier: AAT BIOQUEST INC
22971
76483-836EA 375.27 USD InStock
76483-836
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*, AAT Bioquest
Assays Cellular Assays
Mitochondria are major producers of cellular superoxide.
  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimized for microplate readers but suitable for all major fluorescence platforms
  • results with only 1 hour incubation
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty