LysoBrite™ Red DND-99, AAT Bioquest
Supplier: AAT BIOQUEST INC
22647
76485-128EA 188.87 USD InStock
76485-128
LysoBrite™ Red DND-99, AAT Bioquest
Assays Cellular Assays
LysoBrite Red DND-99 is chemically same to the LysoTracker® Red DND-99 used for labeling and tracking acidic organelles in live cells (LysoTracker® is the trademark of ThermoFisher).
  • Lysosome-Specific Dye
  • Long-Term Use
  • Low Cytotoxicity
  • Stable After Fixation
Precaución: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty