Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Violet 610 succinimidyl ester, mFluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1156
76485-558EA 237.74 USD InStock
76485-558
Violet 610 succinimidyl ester, mFluor™ fluorescent probe
Violet 610 succinimidyl ester
Advances in spectral flow cytometers have expanded applications and capabilities beyond conventional flow cytometry.

  • pH-insensitive
  • High photostability
  • Brighter signal


MW: 1293.46 g/mol
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49293201ec06d5146aeb920fde7c2f612502ef18&vid=49293201ec06d5146aeb920fde7c2f612502ef18&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty