NEBuffer™ Set (r1.1, r2.1, r3.1 and rCutSmart™), New England Biolabs
Supplier: W.u.G. Dickgießer GmbH
B7030S
76486-070EA 29 USD InStock
76486-070
NEBuffer™ Set (r1.1, r2.1, r3.1 and rCutSmart™), New England Biolabs
Buffers Cloning Buffers
New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. This set contains NEBuffers that contain recombinant Albumin.
  • Contains recombinant albumin
  • Set contains 1.25 ml vial of each buffer
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty