UV-Visible Spectrophotometers, Evolution Series, Thermo Scientific™
Supplier: Thermo Scientific
840-341400 840-370100 840-370200 840-341500 840-370400 840-370300 840-340200 840-340100 912A1110 912A1111 840-372600 840-372700
76501-282EA 9795.78 USD InStock
76501-282 76501-284 76501-286 76501-288 76501-290 76501-292 76501-294 76501-296 76501-298 76501-300 76501-308 76501-310
UV-Visible Spectrophotometers, Evolution Series, Thermo Scientific™
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Empower the analysis and bring versatility and productivity to your lab with the Evolution UV-Vis spectrophotometers. With next-generation INSIGHT™ PRO software, the instrument is always up to date and ready for the next sample challenge.
  • Double-beam geometry
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices to complete the analysis
For routine to research analysis, this instrument series offers real innovationsMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty