Laboratory Bottles, Narrow or Wide Neck, Borosilicate Glass, Clear or Amber, with Screw Cap, S.C.A.T.
Supplier: SCAT AMERICAS, INC.
101994 101995 101996 101997 101998 107509 199011 501110 501112 501113 501115 501116 501117 501118 501119 501120 501121 501122 501125 501126 501127 501130 501131 501150 501151 501152 501154 501156 501157 501158
76502-856EA 69.53 USD InStock
76502-856 76502-858 76502-860 76502-862 76502-864 76503-196 76503-720 76504-016 76504-018 76504-020 76504-022 76504-024 76504-026 76504-028 76504-030 76504-032 76504-034 76504-036 76504-038 76504-040 76504-042 76504-044 76504-046 76504-048 76504-050 76504-052 76504-054 76504-056 76504-058 76504-060
Laboratory Bottles, Narrow or Wide Neck, Borosilicate Glass, Clear or Amber, with Screw Cap, S.C.A.T.
Bottles Glass Bottles
Bottles, made of DURAN® borosilicate glass, with blue PP screw cap. Ideal for mixing and storage of culture medium.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Bottles 76504-044 and 76504-046 feature a pour ring to facilitate use.
Información accesorios: Bottle tag multipack consists of (2) blue, (2) purple, (2) green, (2) red, (2) orange, (2) yellow, (2) black, and (2) white for a total of 16 pieces.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty