RNA-Solv® RNA Isolation, Reagent, Omega bio-tek
Supplier: Bierbaum-Proenen GmbH & Co.KG
R6830-02
76509-254EA 298.32 USD InStock
76509-254
RNA-Solv® RNA Isolation, Reagent, Omega bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
RNA-Solv® Reagent is a one reagent system for the isolation of total RNA from cells and tissues. The reagent, a single-phase solution consisting of phenol and guanidine isothiocyanate, is a modification of the single-step RNA isolation method developed by Chomczynski and Sacchi. The sample is homogenised and lysed in RNA-Solv® Reagent, which maintains the integrity of the RNA while disrupting and denaturing endogenous RNases and other cellular components. Extraction of the lysate with chloroform further denatures proteins and separates the mixture into an organic and an aqueous phase. RNA remains exclusively in the aqueous phase, and is subsequently recovered by isopropanol.
This method is suitable for a wide range of starting material: Up to 1 g of tissue or 1×10⁸ cells of human, animal, plant, or bacterial origin. The simplicity of the RNA-Solv® Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in one hour. Total RNA prepared can be used for Northern blot analysis, dot blot hybridization, poly(A)+ selection, in vitro translation, RNase protection assay, and molecular cloning. For use in amplification by thermal cycling, treatment of the isolated RNA with RNase-free DNase I is recommended when the two amplimers lie within a single exon.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty