Deep Well Storage Plates, 96 Round Well, Irish Life Sciences
Supplier: Irish Life Sciences
1.2R96-009U 1.2R96-009US 1.2R96-010UNR 2R96-007U 2R96-007US R96-300V 1R96-304ULP 1R96-322VLP 1.1R96-014ULP 1.1R96-014ULPS
76509-398CS 397.92 USD InStock
76509-398 76509-400 76509-402 76509-404 76509-406 76509-408 76509-410 76509-412 76509-414 76509-416
Deep Well Storage Plates, 96 Round Well, Irish Life Sciences
Microplates Deep Well Microplates
Round well plates made from medical grade polypropylene. Ideal for storage and screening applications.
  • U and V bottom well format for optimal sample recovery
  • Alphanumeric well coding
  • High temperature and chemical resistance (–80 to +121 °C)
  • Autoclavable
  • RNase, DNase Free
  • Stackable for use in plate and automation hotels
This plate has been developed precisely to conform to the footprint dimensions defined by SLAS/ANSI standards. This guarantees its compatibility with both manual and automated sample handling systems, readers, and microplate washers.
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty