Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Compact Shakers, 220 V

Supplier: Yamato Scientific
MK-161-220V MK-201D-220V
76550-268EA 3204.15 USD InStock
76550-268 76550-272
Compact Shakers, 220 V
Shakers and Mixers Rotating/Rocking Shakers
The compact design of this shaker saves precious space.

  • 220 V model
  • Rotary shaking and reciprocate range: 30 mm
  • Various shaking modes included
  • Shaking frequency: 20 to ~200 rpm, random adjustment
  • Digital display of shaking frequency
  • Equipped with shaking timer

It features changeable rotary and reciprocate motions to mix, extract, and stir samples.

A shaking timer is provided, and a digital display indicates shaking frequency. The unit works with IN602CSW incubator for shaking incubation.

Accessories are easy to add and remove.

Precaución: 220More Product Information
Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty