Walk-in-chambers for Stability Testing and Storage, WIC
Supplier: Binder
9800-0001 9800-0002 9800-0003 9800-0004 9800-0007 9800-0008
77041-336EA 79645.93 USD InStock
77041-336 77041-338 77041-340 77041-342 77041-344 77041-346
Walk-in-chambers for Stability Testing and Storage, WIC
Testing Chambers Climatic Chambers
The walk-in-chambers offer a wide range of features and benefits to perfectly meet requirements in stability testing. Perfect test conditions are maintained over a wide range of temperatures from 10 to 50 °C and relative humidity of 20 to 90% RH.
  • Stable and uniform environment
  • Fully compliant with pharma regulations (ICH) equipment
  • Longevity - corrosion resistant
  • Ease of operation
  • Environmentally friendly refrigerant with GWP value 547
  • Maintenance can be carried out with the least possible disruption to the climate conditioning
  • Perfectly meet requirements
More Product Information
Order Now
ORDER
SPECIFICATIONS
SERVICIOS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty