Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromopyrazine 98%

Supplier: AMBEED, INC
Warning

A170269-5G A170269-1G A170269-500G A170269-25G A170269-5G
103844-736EA 10 USD InStock
103844-736 76813-184 76813-186 77081-842 77081-844
2-Amino-5-bromopyrazine 98%
2-Amino-5-bromopyrazine
Precaución: for Research Use Only


Formula: C₄H₄BrN₃
MW: 174.00 g/mol
Boiling Pt: 274.2 °C (760 mm Hg)
Melting Pt: 113…117 °C
Flash Pt: 119.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00235015
CAS Number: 59489-71-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty