OPTI-LYNX™ Holder Tube for 15 mm Guard Cartridge
Supplier: Optimize
11-02328-AA
77100-360EA 115.44 USD InStock
77100-360
OPTI-LYNX™ Holder Tube for 15 mm Guard Cartridge
Chromatography Supplies Chromatography Spare Parts
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
  • For use with OPTI-LYNX™ hardware
  • Installs into holder cap to make connections with guard cartridges
  • Modifies an OPTI-LYNX™ holder to accept guard cartridges
Installs into holder cap to make connections with OPTI-LYNX™ guard column cartridges (15 mm bed length).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty