Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Bromoethynyl)triisopropylsilane 98%

Supplier: AMBEED, INC
Warning

A320630-100G A320630-1G A320630-250MG A320630-25G A320630-5G
77251-128EA 243 USD InStock
77251-128 77251-130 77251-132 77251-134 77251-136 77251-138
(Bromoethynyl)triisopropylsilane 98%
(Bromoethynyl)triisopropylsilane
Precaución: for Research Use Only


Formula: C₁₁H₂₁BrSi
MW: 261.28 g/mol
MDL Number: MFCD11036302
CAS Number: 111409-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty