Standard Spring Clip for SS LocBoard®, ¹/₄" to ¹/₂" Hold Range
Supplier: TRITON PRODUCTS TE
V63205
77269-798PK 20.04 USD InStock
77269-798
Standard Spring Clip for SS LocBoard®, ¹/₄" to ¹/₂" Hold Range
Furniture Backsplashes, Wallracks, Wallpanels
The stainless steel LocHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
  • Ideal for round items such as writing utensils, screwdrivers, box cutters, spray cans, flashlights and more
  • Easily repositioned to accommodate changing storage needs with no damage to the stainless steel LocBoard®
  • Won't rust or corrode in any environment, even ones with extreme temperatures
  • Use multiple hooks for large, heavy and odd-shaped items
  • Vinyl
More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty