Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzoyl chloride 99%

Supplier: AMBEED, INC
Danger

A330739-25G
77290-438EA 26 USD InStock
77290-438
4-Fluorobenzoyl chloride 99%
4-Fluorobenzoyl chloride
Precaución: for Research Use Only


Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 180
Melting Pt: 9
MDL Number: MFCD00000684
CAS Number: 403-43-0
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty