Precision Balances, Legal For Trade, Advanced MX
Supplier: Mettler Toledo
30665177 30665180 30665183 30665186 30665150 30665153 30665156 30665159 30665162 30665165 30665168 30665171 30665174
77323-098EA 7031.58 USD InStock
77323-098 77323-102 77323-106 77323-110 77323-062 77323-066 77323-070 77323-074 77323-078 77323-082 77323-086 77323-090 77323-094
Precision Balances, Legal For Trade, Advanced MX
Balances and Scales
METTLER TOLEDO Advanced MX Legal for Trade Precision Balances feature a precise weigh cell and automatic internal adjustment. The SmartPan stabilizes in any weighing environment, ensuring consistently accurate results.
  • Statistical data analysis
  • Leveling assistant
  • StatusLight
  • 3 interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
Exceptional user management within the 7" touchscreen allows easy access to 9 built-in weighing applications, assigns user access rights and provides automatic data logging. These balances are designed withMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty