Fluorescein-12-dATP *1 mM in Tris Buffer (pH 7.5)*
Supplier: AAT BIOQUEST INC
17036
77461-520EA 106.26 USD InStock
77461-520
Fluorescein-12-dATP *1 mM in Tris Buffer (pH 7.5)*
Nucleic Acid Reagents Nucleotides
Fluorescein-12-dATP might be incorporated into a DNA sequence as its natural counterpart dATP to incorporate a fluorescein tag in a DNA molecule.
  • Can be used for the direct enzymatic labeling of DNA/cDNA, e.g., by PCR or Nick Translation
The linker between fluorescein tag and adenine has been optimized for the optimal substrate incorporation and labeling efficiency.
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty