Orbital Shaker, CO₂ Resistant
Supplier: NEST SCIENTIFIC
105008
77507-388CS 3321.13 USD InStock
77507-388
Orbital Shaker, CO₂ Resistant
Shakers and Mixers Orbital Shakers
This product is developed for it to be used in high humidity and CO₂ environment. Also, it can be used as a common shaker. Its driven mechanism is composed of magnets and sealed electrical coils. It is maintenance free and has long life time.
  • Orbital shaking through magnetic power eliminates the need to replace belts
  • Small footprint ideal for use in a CO₂ incubator
  • Specially treated mechanical components protect up to 20% CO₂ and 95% humidity conditions
  • Minimal heat dissipation assures conditions within the CO₂ chamber are not effected
  • CO₂ resistant shaker
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty