Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gram′s safranin solution

Supplier: G-BIOSCIENCES

786-2099
77540-256EA 254.72 USD InStock
77540-256
Gram′s safranin solution
Gram′s safranin solution
Gram Safranin acts as a counterstain .

  • An aqueous dye solution

It is used in Gram staining to differentiate gram-positive from gram-negative bacteria


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
MDL Number: MFCD00011759
CAS Number: 477-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty