Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amberlyst® 15 (H) H+≥4,7 mmol/g, dry

Supplier: AMBEED, INC
Warning

A920554-1000G A920554-500G
77569-642EA 113 USD InStock
77569-642 77595-484
Amberlyst® 15 (H) H+≥4,7 mmol/g, dry
Amberlyst® 15 (H)
Precaución: For research use only.


Formula: (C₁₀H₁₀·C₈H₈O₃S)x
MDL Number: MFCD00145841
CAS Number: 39389-20-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty