Saddle Vent® Ventilation Kits, Air Systems
Supplier: Air Systems
SVB-E8SCUP
77776-588EA 2416.09 USD InStock
77776-588
Saddle Vent® Ventilation Kits, Air Systems
Air Purifiers
The Saddle Vent® was invented and developed by Air Systems and has been used around the world for over 20 years to make it easier and safer for workers to enter and exit a confined space.
  • Fresh air conduit
  • Only three inch manhole coverage
  • Unrestricted entry and exit
  • Saddle vent and elbow, six and 15 ft. duct, duct canister, universal mount included in kit
The Saddle Vent® takes up only three inches in a manhole opening and functions as a fresh air conduit to the workers in the confined space.
With the Saddle Vent in place, continuous ventilation isMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty