PBS, Phosphate Buffered Saline Tablets, MilliporeSigma
Supplier: EMD MILLIPORE (BIOSCIENCES)
524650-1EA
80058-556EA 120.03 USD InStock
80058-556
PBS, Phosphate Buffered Saline Tablets, MilliporeSigma
Buffers Electrophoresis Buffers Washing Buffers
Dissolving one tablet in 1 liter of deionized H₂O yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7.4 at 25°C.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty