Spatulas, Spoon, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
Supplier: Chemglass
CG-1983-12
80071-672EA 9.14 USD InStock
80071-672
Spatulas, Spoon, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
Spatulas
Spoon-type spatula is made of non-magnetic stainless steel with an overall length of 6.5" (165 mm).
  • Spatula will pass through a 14/10 or 14/20 outer joint
  • The oval spoon end is ³/₄" (19 mm) long × ³/₁₆" (4.8 mm) wide
  • Equipped with a cushion grip vinyl handle for easy holding
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty