Galvanic Oxygen Sensors, WTW
Supplier: WTW
201425 201533 201545 201546 201548 201549 201570
80085-410EA 2770 USD InStock
80085-410 80085-412 80085-420 80085-422 80085-424 80085-426 80085-428 80085-430 80085-432
Galvanic Oxygen Sensors, WTW
Electrodes Dissolved Oxygen Electrodes
These galvanic sensors are ready-to-use and are equipped with protective hoods for rough conditions.
CellOx* sensors are membrane-covered and waterproof. The membrane can be used for up to six months. Sensor also features high signal resolution, temperature compensation, rapid response, and membrane leakage monitoring. Includes a waterproof plug.
DurOx* oxygen sensors are membrane-covered and waterproof. Use for up to six months with only one electrolyte filling. Features temperature compensation and low approach flow. Includes a waterproof plug.
The StirrOx* self-stirring oxygenMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty